9-(4-Bromophenyl)-9-phenylfluorene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 9-(4-Bromophenyl)-9-phenyl-9H-fluorene

B5221-1G B5221-5G
TCIAB5221-1GEA 535 SEK
TCIAB5221-1G TCIAB5221-5G
9-(4-Bromophenyl)-9-phenylfluorene ≥98.0%
9-(4-Bromophenyl)-9-phenylfluorene
Formel: C₂₅H₁₇Br
MW: 397,32 g/mol
MDL Number: MFCD28975087
CAS nummer: 937082-81-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR