4'-Butyl-4-iodobiphenyl ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 4-Butyl-4'-iodobiphenyl
B5211-200MG B5211-1G
TCIAB5211-200MGEA 455 SEK
TCIAB5211-200MG TCIAB5211-1G
4'-Butyl-4-iodobiphenyl ≥98.0%
4'-Butyl-4-iodobiphenyl
Formel: C₁₆H₁₇I
MW: 336,22 g/mol
MDL Number: MFCD16619281
CAS nummer: 199982-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59b49b2d8a58d0369a515126397d6ada20a1f507&vid=59b49b2d8a58d0369a515126397d6ada20a1f507&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 122.0 deg-C