2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene ≥98.0%
Leverantör: TCI EUROPE
B5196-200MG
B5196-1G
TCIAB5196-200MGEA
1600
SEK
TCIAB5196-200MG
TCIAB5196-1G
2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene ≥98.0%
2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene
Formel:
C₁₆H₂₀S₂Si₂ MW: 332,63 g/mol |
CAS nummer:
139896-65-4 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 211.0 to 215.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...