2-Bromoisonicotinonitrile ≥98.0%

Leverantör: TCI
Fara

B5189-1G B5189-5G
TCIAB5189-1GEA 670 SEK
TCIAB5189-1G TCIAB5189-5G
2-Bromoisonicotinonitrile ≥98.0%
2-Bromoisonicotinonitrile
Formel: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD07367879
CAS nummer: 10386-27-3
UN: 3439
ADR: 6.1 III

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR