9-(Bromomethyl)nonadecane ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
B5188-5ML
B5188-25ML
TCIAB5188-5MLEA
1100
SEK
TCIAB5188-5ML
TCIAB5188-25ML
9-(Bromomethyl)nonadecane ≥96.0%
9-(Bromomethyl)nonadecane
Formel:
C₂₀H₄₁Br MW: 361,45 g/mol Kokpunkt: 195°C |
MDL Number:
MFCD28515435 CAS nummer: 69620-20-8 |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...