9-(3-Bromopropyl)carbazole ≥98.0%

Leverantör: TCI
Varning

B5183-1G B5183-200MG
TCIAB5183-1GEA 4150 SEK
TCIAB5183-1G TCIAB5183-200MG
9-(3-Bromopropyl)carbazole ≥98.0%
9-(3-Bromopropyl)carbazole
Formel: C₁₅H₁₄BrN
MW: 288.19 g/mol
MDL Number: MFCD11164489
CAS nummer: 84359-61-5

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR