4-Bromo-3-phenylpyrazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4-Bromo-3-phenyl-1H-pyrazole

B5181-200MG B5181-1G
TCIAB5181-200MGEA 510 SEK
TCIAB5181-200MG TCIAB5181-1G
4-Bromo-3-phenylpyrazole ≥98.0%
4-Bromo-3-phenylpyrazole
Formel: C₉H₇BrN₂
MW: 223.07 g/mol
MDL Number: MFCD09027561
CAS nummer: 13808-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3428be08aec1dd8b500f300212915d2008d52a45&vid=3428be08aec1dd8b500f300212915d2008d52a45&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR