3-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
|
Varning
|
B5178-5G
B5178-25G
TCIAB5178-5GEA
335
SEK
TCIAB5178-5G
TCIAB5178-25G
3-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
3-Bromobenzamide
Formel:
C₇H₆BrNO |
MDL Number:
MFCD00017127 CAS nummer: 22726-00-7 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...