4-Benzyloxy-3-methylphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: (4-(Benzyloxy)-3-methylphenyl)boronic acid, 4-Benzyloxy-3-methylbenzeneboronic acid

B5169-1G B5169-5G
TCIAB5169-1GEA 845 SEK
TCIAB5169-1G TCIAB5169-5G
4-Benzyloxy-3-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-3-methylphenylboronic acid
Formel: C₁₄H₁₅BO₃
Smältpunkt: 130°C
MDL Number: MFCD06409944
CAS nummer: 338454-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72dbf241eee3e0f26d8e188d89a8e9aebd2259f4&vid=72dbf241eee3e0f26d8e188d89a8e9aebd2259f4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR