3-Bromo-5-chloro-2-methoxypyridine ≥98.0%
Leverantör: TCI
|
Varning
|
B5165-1G
B5165-5G
TCIAB5165-1GEA
895
SEK
TCIAB5165-1G
TCIAB5165-5G
3-Bromo-5-chloro-2-methoxypyridine ≥98.0%
3-Bromo-5-chloro-2-methoxypyridine
Formel:
C₆H₅BrClNO |
MDL Number:
MFCD08059322 CAS nummer: 102830-75-1 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...