tert-Butyldimethyl(2-propynyloxy)silane ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Tert-Butyldimethyl(2-propynyloxy)silane, tert-Butyldimethyl(prop-2-ynyloxy)silane
B5163-5ML B5163-25ML
TCIAB5163-5MLEA 670 SEK
TCIAB5163-5ML TCIAB5163-25ML
tert-Butyldimethyl(2-propynyloxy)silane ≥98.0%
tert-Butyldimethyl(2-propynyloxy)silane
Formel: C₉H₁₈OSi
MW: 170,33 g/mol
Kokpunkt: 163°C
MDL Number: MFCD28156589
CAS nummer: 76782-82-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e61c02493e6fc763d8fe775aebaf1eca3807041&vid=7e61c02493e6fc763d8fe775aebaf1eca3807041&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %