3-(2-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
B5152-200MG
B5152-1G
TCIAB5152-200MGEA
1150
SEK
TCIAB5152-200MG
TCIAB5152-1G
3-(2-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
3-(2-Bromophenyl)pyridine
Formel:
C₁₁H₈BrN MW: 234,10 g/mol Kokpunkt: 191°C |
MDL Number:
MFCD18415514 CAS nummer: 847264-30-6 |
Specifikation testresultat
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...