(2-Bromophenylethynyl)trimethylsilane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: ((2-Bromophenyl)ethynyl)trimethylsilane

B5138-1G B5138-5G
TCIAB5138-1GEA 390 SEK
TCIAB5138-1G TCIAB5138-5G
(2-Bromophenylethynyl)trimethylsilane ≥98.0%
(2-Bromophenylethynyl)trimethylsilane
Formel: C₁₁H₁₃BrSi
MW: 253,21 g/mol
Kokpunkt: 91°C
Storage Temperature: Kylskåp
MDL Number: MFCD01321391
CAS nummer: 38274-16-7

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR