3-Bromo-9-(4-bromophenyl)-9H-carbazole ≥97.0%

Leverantör: TCI EUROPE
Varning

B5107-1G B5107-5G
TCIAB5107-1GEA 1375 SEK
TCIAB5107-1G TCIAB5107-5G
3-Bromo-9-(4-bromophenyl)-9H-carbazole ≥97.0%
3-Bromo-9-(4-bromophenyl)-9H-carbazole
Formel: C₁₈H₁₁Br₂N
MW: 401,10 g/mol
MDL Number: MFCD30063199
CAS nummer: 1226860-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5163912e1b1e8823e09a76e20a84a002986f4527&vid=5163912e1b1e8823e09a76e20a84a002986f4527&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR