1-Bromo-4-phenylnaphthalene ≥98.0%

Leverantör: TCI

B5105-1G B5105-5G
TCIAB5105-1GEA 1300 SEK
TCIAB5105-1G TCIAB5105-5G
1-Bromo-4-phenylnaphthalene ≥98.0%
1-Bromo-4-phenylnaphthalene
Formel: C₁₆H₁₁Br
Kokpunkt: 153°C
MDL Number: MFCD17012346
CAS nummer: 59951-65-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR