ω-Bromo-2-fluoroacetophenone ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Bromomethyl o-fluorophenyl ketone, 2-Bromo-2'-fluoroacetophenone, α-Bromo-2-fluoroacetophenone, 2-Fluorophenacyl bromide, 2-Bromo-1-(2-fluorophenyl)ethan-1-one, 2-Bromo-1-(2-fluorophenyl)ethanone

B5104-5G B5104-25G
TCIAB5104-5GEA 1250 SEK
TCIAB5104-5G TCIAB5104-25G
ω-Bromo-2-fluoroacetophenone ≥98.0%
ω-Bromo-2-fluoroacetophenone
Formel: C₈H₆BrFO
MW: 217,04 g/mol
Kokpunkt: 63°C
Storage Temperature: Kylskåp
MDL Number: MFCD00278796
CAS nummer: 655-15-2
UN: 3261
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR