4-Bromo-3-nitrobenzaldehyde ≥98.0%

Leverantör: TCI
Varning

B5101-1G B5101-5G
TCIAB5101-1GEA 405 SEK
TCIAB5101-1G TCIAB5101-5G
4-Bromo-3-nitrobenzaldehyde ≥98.0%
4-Bromo-3-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
MDL Number: MFCD00204031
CAS nummer: 163596-75-6

Order Now

Specifikation testresultat

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR