4-Bromo-1-ethyl-1H-pyrazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Bromo-1-ethylpyrazole
B5100-1G
B5100-5G
TCIAB5100-1GEA
655
SEK
TCIAB5100-1G
TCIAB5100-5G
4-Bromo-1-ethyl-1H-pyrazole ≥98.0%
4-Bromo-1-ethyl-1H-pyrazole
Formel:
C₅H₇BrN₂ MW: 175,0265 g/mol Kokpunkt: 108°C |
MDL Number:
MFCD08235252 CAS nummer: 71229-85-1 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...