7-Bromooxindole ≥98.0%

Leverantör: TCI EUROPE

B5097-200MG B5097-1G
TCIAB5097-200MGEA 550 SEK
TCIAB5097-200MG TCIAB5097-1G
7-Bromooxindole ≥98.0%
7-Bromooxindole
Formel: C₈H₆BrNO
MW: 212,0433 g/mol
MDL Number: MFCD02179613
CAS nummer: 320734-35-8

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 210.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR