4-Bromo-2-methylindan-1-one ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4-Bromo-2-methyl-2,3-dihydro-1H-inden-1-one, 4-Bromo-2-methyl-1-indanone

B5096-1G B5096-5G
TCIAB5096-1GEA 1075 SEK
TCIAB5096-1G TCIAB5096-5G
4-Bromo-2-methylindan-1-one ≥98.0%
4-Bromo-2-methylindan-1-one
Formel: C₁₀H₉BrO
Kokpunkt: 134°C
MDL Number: MFCD09881709
CAS nummer: 174702-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2da5a702b15354536ef7698c6ce43ea556636ed9&vid=2da5a702b15354536ef7698c6ce43ea556636ed9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR