3-Bromo-4-ethoxybenzaldehyde ≥97.0%

Leverantör: TCI EUROPE
Varning

B5079-1G B5079-5G
TCIAB5079-1GEA 485 SEK
TCIAB5079-1G TCIAB5079-5G
3-Bromo-4-ethoxybenzaldehyde ≥97.0%
3-Bromo-4-ethoxybenzaldehyde
Formel: C₉H₉BrO₂
MW: 229,07 g/mol
MDL Number: MFCD00672941
CAS nummer: 108373-05-3

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR