5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene ≥98.0%

Leverantör: TCI EUROPE
Varning

B5057-1G B5057-5G
TCIAB5057-1GEA 1125 SEK
TCIAB5057-1G TCIAB5057-5G
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene ≥98.0%
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Formel: C₇H₂BrF₅O
MW: 276,99 g/mol
MDL Number: MFCD03412225
CAS nummer: 115467-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14afc6199843248904aab89bc0b63abf5a45f227&vid=14afc6199843248904aab89bc0b63abf5a45f227&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR