1-Bromo-4-(4-bromophenyl)naphthalene ≥97.0%
Leverantör: TCI EUROPE
B5056-1G
B5056-5G
TCIAB5056-1GEA
1975
SEK
TCIAB5056-1G
TCIAB5056-5G
1-Bromo-4-(4-bromophenyl)naphthalene ≥97.0%
1-Bromo-4-(4-bromophenyl)naphthalene
Formel:
C₁₆H₁₀Br₂ MW: 362,06 g/mol |
CAS nummer:
952604-26-1 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...