Bis(3-biphenylyl)amine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI
Varning

Synonyms: 3,3'-Iminobis(biphenyl), Di([1,1'-biphenyl]-3-yl)amine

B5048-200MG B5048-1G
TCIAB5048-200MGEA 1350 SEK
TCIAB5048-200MG TCIAB5048-1G
Bis(3-biphenylyl)amine ≥98.0% (by HPLC, total nitrogen)
Bis(3-biphenylyl)amine
Formel: C₂₄H₁₉N
MW: 321.42 g/mol
MDL Number: MFCD28129440
CAS nummer: 169224-65-1

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR