4'-Bromotri(4-biphenylyl)amine ≥98.0%

Leverantör: TCI

Synonyms: N,N-Di(4-biphenylyl)-N-(4'-bromo-4-biphenylyl)amine, N,N-Di([1,1'-biphenyl]-4-yl)-4'-bromo-[1,1'-biphenyl]-4-amine

B5020-1G B5020-5G
TCIAB5020-1GEA 1150 SEK
TCIAB5020-1G TCIAB5020-5G
4'-Bromotri(4-biphenylyl)amine ≥98.0%
4'-Bromotri(4-biphenylyl)amine
Formel: C₃₆H₂₆BrN
MW: 552.52 g/mol
MDL Number: MFCD22571699
CAS nummer: 728039-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f8250774251c80c4425847a20dacd654f1101ea&vid=6f8250774251c80c4425847a20dacd654f1101ea&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 225.0 to 229.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR