2-(4-Bromophenyl)benzo[b]thiophene ≥98.0%
Leverantör: TCI EUROPE
B5016-200MG
B5016-1G
TCIAB5016-200MGEA
855
SEK
TCIAB5016-200MG
TCIAB5016-1G
2-(4-Bromophenyl)benzo[b]thiophene ≥98.0%
2-(4-Bromophenyl)benzo[b]thiophene
Formel:
C₁₄H₉BrS MW: 289,19 g/mol |
MDL Number:
MFCD22380322 CAS nummer: 19437-86-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 213.0 to 217.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...