3-Bromo-5-ethoxypyridine ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
B5000-1G
B5000-5G
TCIAB5000-1GEA
910
SEK
TCIAB5000-1G
TCIAB5000-5G
3-Bromo-5-ethoxypyridine ≥97.0% (by GC, titration analysis)
3-Bromo-5-ethoxypyridine
Formel:
C₇H₈BrNO MW: 202,05 g/mol Kokpunkt: 240°C Smältpunkt: 9°C |
MDL Number:
MFCD00234116 CAS nummer: 17117-17-8 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...