N-(6-Bromohexyl)phthalimide ≥98.0%

Leverantör: TCI EUROPE

B4987-5G B4987-25G
TCIAB4987-5GEA 605 SEK
TCIAB4987-5G TCIAB4987-25G
N-(6-Bromohexyl)phthalimide ≥98.0%
N-(6-Bromohexyl)phthalimide
Formel: C₁₄H₁₆BrNO₂
MW: 310,19 g/mol
Kokpunkt: 188°C
MDL Number: MFCD00023098
CAS nummer: 24566-79-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR