1-tert-Butylimidazole ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

B4985-5G B4985-25G
TCIAB4985-5GEA 2225 SEK
TCIAB4985-5G TCIAB4985-25G
1-tert-Butylimidazole ≥98.0% (by GC, titration analysis)
1-tert-Butylimidazole
Formel: C₇H₁₂N₂
MW: 124,19 g/mol
Kokpunkt: 105°C
MDL Number: MFCD19441219
CAS nummer: 45676-04-8

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR