4-tert-Butylphenyl isothiocyanate ≥98.0%
Leverantör: TCI
Fara
Synonyms: 4-tert-Butylphenylisothiocyanate
B4984-1G B4984-5G
TCIAB4984-1GEA 375 SEK
TCIAB4984-1G TCIAB4984-5G
4-tert-Butylphenyl isothiocyanate ≥98.0%
4-tert-Butylphenyl isothiocyanate
Formel: C₁₁H₁₃NS
Kokpunkt: 277°C
Storage Temperature: Kylskåp
MDL Number: MFCD00022059
CAS nummer: 19241-24-8
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C