3-Bromo-5-(trifluoromethyl)benzaldehyde ≥98.0%
Leverantör: TCI
Varning
Synonyms: 3-Bromo-5-trifluoromethylbenzaldehyde
B4974-1G B4974-5G
TCIAB4974-1GEA 310 SEK
TCIAB4974-1G TCIAB4974-5G
3-Bromo-5-(trifluoromethyl)benzaldehyde ≥98.0%
3-Bromo-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₄BrF₃O
MW: 253.02 g/mol
MDL Number: MFCD08059506
CAS nummer: 477535-41-4
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %