Benzo[b]naphtho[1,2-d]thiophene ≥98.0%
Leverantör: TCI EUROPE
B4951-1G B4951-5G
TCIAB4951-1GEA 1225 SEK
TCIAB4951-1G TCIAB4951-5G
Benzo[b]naphtho[1,2-d]thiophene ≥98.0%
Benzo[b]naphtho[1,2-d]thiophene
Formel: C₁₆H₁₀S
MW: 234,32 g/mol
Kokpunkt: 220°C
Storage Temperature: Kylskåp
MDL Number: MFCD00215942
CAS nummer: 205-43-6
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C