4-Bromo-1-naphthoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Bromo-1-naphthalenecarboxylic acid
B4949-1G
B4949-5G
TCIAB4949-1GEA
350
SEK
TCIAB4949-1G
TCIAB4949-5G
4-Bromo-1-naphthoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-1-naphthoic acid
Formel:
C₁₁H₇BrO₂ MW: 251,08 g/mol |
MDL Number:
MFCD00094221 CAS nummer: 16650-55-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 223.0 to 227.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...