2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine ≥97.0% (by GC, total nitrogen)

Leverantör: TCI
Varning

B4940-200MG B4940-1G
TCIAB4940-200MGEA 390 SEK
TCIAB4940-200MG TCIAB4940-1G
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine ≥97.0% (by GC, total nitrogen)
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine
Formel: C₁₅H₉Cl₂N₃
MW: 302.16 g/mol
MDL Number: MFCD28556865
CAS nummer: 10202-45-6

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (with Total Nitrogen) min. 97.0 %
Melting point 157.0 to 161.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR