2-Benzoylpyrrole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Phenyl(1H-pyrrol-2-yl)methanone

B4934-5G B4934-25G
TCIAB4934-5GEA 1400 SEK
TCIAB4934-5G TCIAB4934-25G
2-Benzoylpyrrole ≥98.0%
2-Benzoylpyrrole
Formel: C₁₁H₉NO
MW: 171,20 g/mol
Kokpunkt: 305°C
MDL Number: MFCD00094048
CAS nummer: 7697-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70516ddaaede638d56d7908fee742bd81a606bb8&vid=70516ddaaede638d56d7908fee742bd81a606bb8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR