2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

B4927-1G B4927-5G
TCIAB4927-1GEA 950 SEK
TCIAB4927-1G TCIAB4927-5G
2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine
Formel: C₂₁H₁₄BrN₃
MW: 388,27 g/mol
MDL Number: MFCD00194632
CAS nummer: 23449-08-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 200.0 to 204.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR