4-Butyl-4'-cyanobiphenyl ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4'-Butyl-[1,1'-biphenyl]-4-carbonitrile
B4923-5G B4923-25G
TCIAB4923-5GEA 630 SEK
TCIAB4923-5G TCIAB4923-25G
4-Butyl-4'-cyanobiphenyl ≥98.0%
4-Butyl-4'-cyanobiphenyl
Formel: C₁₇H₁₇N
MW: 235,33 g/mol
Kokpunkt: 195°C
MDL Number: MFCD00506009
CAS nummer: 52709-83-8
EINECS: 258-119-0
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C
Mesomorphic range 45.0 to 51.0 deg-C