4-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0%

Leverantör: TCI

Synonyms: 4-Bromo-5'-phenyl-m-terphenyl, 5'-(4-Bromophenyl)-1,1':3',1''-terphenyl, 5'-(4-Bromophenyl)-m-terphenyl

B4889-1G B4889-5G
TCIAB4889-1GEA 4900 SEK
TCIAB4889-1G TCIAB4889-5G
4-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0%
4-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formel: C₂₄H₁₇Br
MW: 385.30 g/mol
MDL Number: MFCD26407506
CAS nummer: 116941-52-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR