3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)-[1,1'-biphenyl]-4-amine
B4887-1G
B4887-5G
TCIAB4887-1GEA
870
SEK
TCIAB4887-1G
TCIAB4887-5G
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole ≥98.0% (by HPLC, total nitrogen)
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole
Formel:
C₃₆H₂₆N₂ MW: 486,62 g/mol |
MDL Number:
MFCD22571697 CAS nummer: 1160294-96-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 178.0 to 187.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...