1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: BHFC

B4881-1G B4881-5G
TCIAB4881-1GEA 3825 SEK
TCIAB4881-1G TCIAB4881-5G
1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene ≥97.0%
1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene
Formel: C₉H₇F₇O
MW: 264,14 g/mol
MDL Number: MFCD00084878
CAS nummer: 958032-89-8

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR