4-Bromo-3-nitroaniline ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B4876-1G
B4876-5G
TCIAB4876-1GEA
365
SEK
TCIAB4876-1G
TCIAB4876-5G
4-Bromo-3-nitroaniline ≥98.0%
4-Bromo-3-nitroaniline
Formel:
C₆H₅BrN₂O₂ MW: 217,02 g/mol |
MDL Number:
MFCD01320678 CAS nummer: 53324-38-2 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 131.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...