2-(4-Bromophenyl)imidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
B4871-1G
B4871-5G
TCIAB4871-1GEA
695
SEK
TCIAB4871-1G
TCIAB4871-5G
2-(4-Bromophenyl)imidazo[1,2-a]pyridine ≥98.0% (by GC, titration analysis)
2-(4-Bromophenyl)imidazo[1,2-a]pyridine
Formel:
C₁₃H₉BrN₂ MW: 273,13 g/mol |
MDL Number:
MFCD00218250 CAS nummer: 34658-66-7 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 215.0 to 219.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...