(±)-1-Benzyl-3-hydroxypiperidine ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-Benzyl-3-hydroxypiperidine, 1-Benzylpiperidin-3-ol, 1-Benzyl-3-piperidinol, 1-Benzyl-3-hydroxypiperidine
B4868-5G
B4868-25G
TCIAB4868-5GEA
630
SEK
TCIAB4868-5G
TCIAB4868-25G
(±)-1-Benzyl-3-hydroxypiperidine ≥97.0% (by GC, titration analysis)
(±)-1-Benzyl-3-hydroxypiperidine
Formel:
C₁₂H₁₇NO MW: 191,27 g/mol Kokpunkt: 153°C |
MDL Number:
MFCD00023734 CAS nummer: 14813-01-5 EINECS: 238-881-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...