2-Bromo-8-iododibenzofuran ≥98.0%

Leverantör: TCI

B4867-200MG B4867-1G
TCIAB4867-200MGEA 630 SEK
TCIAB4867-200MG TCIAB4867-1G
2-Bromo-8-iododibenzofuran ≥98.0%
2-Bromo-8-iododibenzofuran
Formel: C₁₂H₆BrIO
MDL Number: MFCD28975101
CAS nummer: 916435-41-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR