N-(4-Biphenylyl)-2-biphenylamine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B4866-1G
B4866-5G
TCIAB4866-1GEA
935
SEK
TCIAB4866-1G
TCIAB4866-5G
N-(4-Biphenylyl)-2-biphenylamine ≥98.0%
N-(4-Biphenylyl)-2-biphenylamine
Formel:
C₂₄H₁₉N MW: 321,42 g/mol |
MDL Number:
MFCD22988894 CAS nummer: 1372775-52-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...