3-(3-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
B4865-1G
B4865-5G
TCIAB4865-1GEA
805
SEK
TCIAB4865-1G
TCIAB4865-5G
3-(3-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
3-(3-Bromophenyl)pyridine
Formel:
C₁₁H₈BrN MW: 234,10 g/mol |
MDL Number:
MFCD16658917 CAS nummer: 4422-32-6 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...