(R)-(-)-1-Benzyl-3-hydroxypiperidine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(R)-1-Benzyl-3-piperidinol
B4857-1G
B4857-5G
TCIAB4857-1GEA
965
SEK
TCIAB4857-1G
TCIAB4857-5G
(R)-(-)-1-Benzyl-3-hydroxypiperidine ≥98.0%
(R)-(-)-1-Benzyl-3-hydroxypiperidine
Formel:
C₁₂H₁₇NO MW: 191,27 g/mol Kokpunkt: 102°C |
MDL Number:
MFCD00274307 CAS nummer: 91599-81-4 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -10.0 to -13.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...