6-Bromo-1-tetralone ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Bromo-3,4-dihydro-1(2H)-naphthalenone, 6-Bromo-3,4-dihydronaphthalen-1(2H)-one
B4841-1G
B4841-5G
TCIAB4841-1GEA
1700
SEK
TCIAB4841-1G
TCIAB4841-5G
6-Bromo-1-tetralone ≥95.0%
6-Bromo-1-tetralone
Formel:
C₁₀H₉BrO MW: 225,08 g/mol Kokpunkt: 116°C |
MDL Number:
MFCD04114378 CAS nummer: 66361-67-9 |
Specifikation testresultat
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 95.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...