7-Bromobenzo[b]thiophene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B4829-200MG
B4829-1G
TCIAB4829-200MGEA
485
SEK
TCIAB4829-200MG
TCIAB4829-1G
7-Bromobenzo[b]thiophene ≥98.0%
7-Bromobenzo[b]thiophene
Formel:
C₈H₅BrS MW: 213,0943 g/mol Kokpunkt: 109°C |
MDL Number:
MFCD06657729 CAS nummer: 1423-61-6 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...