Benorilate ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI

Synonyms: Aspirin Acetaminophen ester, 4-Acetamidophenyl 2-Acetoxybenzoate

B4827-5G B4827-25G
TCIAB4827-5GEA 525 SEK
TCIAB4827-5G TCIAB4827-25G
Benorilate ≥98.0% (by HPLC, total nitrogen)
Benorilate
Formel: C₁₇H₁₅NO₅
MW: 313.31 g/mol
MDL Number: MFCD00864257
CAS nummer: 5003-48-5
EINECS: 225-674-5
Merck Index: 14,01045

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 178.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR